6-(Bromomethyl)quinoxaline 97%

Supplier: Ambeed
Danger

A480670-100MG A480670-250MG
103828-120EA 428 USD
103828-120 77034-340 77034-342
6-(Bromomethyl)quinoxaline 97%
6-(Bromomethyl)quinoxaline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
Boiling Pt: 325.7 °C (760 mm Hg)
Flash Pt: 150.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11559042
CAS Number: 53967-21-8
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR