6-(Benzyloxy)-9-((1S,3S)-4-(benzyloxy)-3-((benzyloxy)methyl)-2-methylenecyclopentyl)-N-((4-methoxyphenyl)diphenylmethyl)-9H-purin-2-amine 95%

Supplier: Ambeed
Warning

A100394-100MG A100394-250MG A100394-1G
103795-416EA 61 USD
103795-416 103821-114 76994-438 76994-440 77296-668
6-(Benzyloxy)-9-((1S,3S)-4-(benzyloxy)-3-((benzyloxy)methyl)-2-methylenecyclopentyl)-N-((4-methoxyphenyl)diphenylmethyl)-9H-purin-2-amine 95%
6-(Benzyloxy)-9-((1S,3S)-4-(benzyloxy)-3-((benzyloxy)methyl)-2-methylenecyclopentyl)-N-((4-methoxyphenyl)diphenylmethyl)-9H-purin-2-amine

Precaución: for Research Use Only

Formula: C₅₃H₄₉N₅O₄
MW: 819.98 g/mol
Storage Temperature: Ambient
CAS Number: 142217-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR