6-(Benzyloxy)-7H-purin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: O6-Benzylguanine

A394978-1G A394978-25G A394978-10G A394978-500G
103826-954EA 5 USD
103826-954 103837-992 76841-414 76841-416 77252-064 77746-790
6-(Benzyloxy)-7H-purin-2-amine 98%
6-(Benzyloxy)-7H-purin-2-amine

Precaución: for Research Use Only

Formula: C12H11N5O
MW: 241.2 g/mol
Flash Pt: 329.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00269931
CAS Number: 19916-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR