6-Quinoxaline​boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: Quinoxaline-6-boronic acid, pinacol ester, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline

A635991-1G A635991-5G A635991-100MG A635991-250MG
103840-314EA 331 USD
103840-314 76844-484 76844-486 77035-360 77035-362
6-Quinoxaline​boronic acid pinacol ester 95%
6-Quinoxaline​boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₇BN₂O₂
MW: 256.1080 g/mol
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11054040
CAS Number: 1167418-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR