(2,3-Dihydro-3-oxo-1H-isoindol-5-yl)boronic acid, pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-isoindol-1-one, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one, 2,3-Dihydro-1-oxo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-isoindole

A181496-100MG A181496-250MG A181496-1G A181496-5G
103807-610EA 83 USD
103807-610 103845-984 77029-444 77029-446 77083-508 77083-510
(2,3-Dihydro-3-oxo-1H-isoindol-5-yl)boronic acid, pinacol ester 95%
(2,3-Dihydro-3-oxo-1H-isoindol-5-yl)boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₃
Boiling Pt: 466.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11849992
CAS Number: 1004294-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR