Oxindole-6-boronic acid pinacol ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-2-one
A154076-100MG
A154076-250MG
A154076-1G
A154076-5G
103804-600EA
48
USD
103804-600
103842-780
76973-246
76973-248
77318-470
77318-472
Oxindole-6-boronic acid pinacol ester 95%
Oxindole-6-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BNO₃ MW: 259.1086 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11855965 CAS Number: 893441-85-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...