6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)imidazo[1,2-a]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: Imidazo[1,2-a]pyridine-6-boronic acid, pinacol ester

A117812-100MG A117812-250MG A117812-1G A117812-5G
103798-938EA 84 USD
103798-938 103824-830 76822-226 76822-228 77311-606 77311-608
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)imidazo[1,2-a]pyridine 95%
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)imidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₇BN₂O₂
MW: 244.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181965
CAS Number: 1204742-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR