6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazin-3(4H)-one 95%

Supplier: Ambeed
Warning

Synonyms: 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester

A239885-100MG A239885-250MG A239885-1G A239885-5G
103811-316EA 123 USD
103811-316 103851-114 77031-262 77031-264 77090-264 77090-266
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazin-3(4H)-one 95%
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C14H18BNO4
Boiling Pt: 441.1 °C (760 mm Hg)
Melting Pt: 190…194 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12755796
CAS Number: 943994-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR