6-Indoleboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
A172947-1G
A172947-25G
A172947-5G
A172947-10G
A172947-250MG
103806-768EA
19
USD
103806-768
103845-060
77082-278
77082-280
77082-282
77111-090
77111-092
6-Indoleboronic acid pinacol ester 97%
6-Indoleboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BNO₂ Boiling Pt: 396 °C (760 mm Hg) Melting Pt: 167…172 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07368852 CAS Number: 642494-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...