6-(3-Methoxyphenyl)-4-methyl-2-(2-morpholinoethyl)pyridazin-3(2H)-one hydrochloride 98%

Supplier: Ambeed
Warning

A1001772-1MG A1001772-10MG A1001772-50MG A1001772-5MG
103791-370EA 28 USD
103791-370 103791-852 103799-206 76645-908 77296-580 77296-582 77296-584
6-(3-Methoxyphenyl)-4-methyl-2-(2-morpholinoethyl)pyridazin-3(2H)-one hydrochloride 98%
6-(3-Methoxyphenyl)-4-methyl-2-(2-morpholinoethyl)pyridazin-3(2H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₄ClN₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD01692601
CAS Number: 13299-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR