6-[3-(Trifluoromethyl)phenoxy]-2-pyridinecarboxylic acid ≥98%

Supplier: Ambeed
Warning

A307508-250MG A307508-5G
103814-506EA 107 USD
103814-506 77672-554 77872-426
6-[3-(Trifluoromethyl)phenoxy]-2-pyridinecarboxylic acid ≥98%
6-[3-(Trifluoromethyl)phenoxy]-2-pyridinecarboxylic acid

Precaución: Sealed in dry, at 2…8°C

Formula: C₁₃H₈F₃NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00227526
CAS Number: 137640-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR