Adapalene 99%

Supplier: Ambeed
Warning

Synonyms: 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic Acid, 6-(3-(Adamantan-1-yl)-4-methoxyphenyl)-2-naphthoic acid

A105293-1MG A105293-250MG A105293-25G A105293-5G A105293-1G
103796-868EA 35 USD
103796-868 76994-846 76994-848 76994-850 76994-852 77309-464
Adapalene 99%
Adapalene

Precaución: for Research Use Only

Formula: C₂₈H₂₈O₃
MW: 412.53 g/mol
Boiling Pt: 606.3 °C (760 mm Hg)
Melting Pt: 319…322 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03106112
CAS Number: 106685-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR