EMCH.TFA (E-Maleimidocaproic acid hydrazine trifluoroacetic acid salt) 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
EMCH, E-Maleimidocaproic acid hydrazine trifluoroacetic acid salt, 3,3'-N-[ε-Maleimidocaproic acid] hydrazide trifluoroacetic acid salt, EMCH.TFA, 6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanehydrazide 2,2,2-trifluoroacetate, 6-Maleimidohexanehydrazide trifluoroacetate, 6-Maleimidocaproic acid hydrazide TFA, N-ε-Maleimidocaproic acid hydrazide trifluoroacetic acid salt
A100170-100MG
A100170-1G
A100170-250MG
103795-274EA
70
USD
103795-274
103820-990
76809-190
76809-192
77282-446
77610-726
EMCH.TFA (E-Maleimidocaproic acid hydrazine trifluoroacetic acid salt) 97%
EMCH.TFA (E-Maleimidocaproic acid hydrazine trifluoroacetic acid salt)
Precaución: for Research Use Only
Formula:
C₁₀H₁₅N₃O₃·C₂HF₃O₂ Melting Pt: 104…105 °C Storage Temperature: Freezer |
MDL Number:
MFCD07357647 CAS Number: 151038-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...