6-(2,2,2-Trifluoroethoxy)nicotinaldehyde 98%

Supplier: Ambeed
Warning

A169201-100MG A169201-10G A169201-250MG A169201-1G
103806-364EA 86 USD
103806-364 103844-618 77011-542 77011-544 77011-546 77300-970
6-(2,2,2-Trifluoroethoxy)nicotinaldehyde 98%
6-(2,2,2-Trifluoroethoxy)nicotinaldehyde

Precaución: for Research Use Only

Formula: C8H6F3NO2
Storage Temperature: Refrigerator
MDL Number: MFCD06738679
CAS Number: 159981-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR