5’-Deoxy-5’-methylthioadenosine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methylthioadenosine, 5'-S-methyl-5'-thio-adenosine, NSC 335422, Vitamil L2, MTA, (2R,3R,4S,5S)-2-(6-Amino-9H-purin-9-yl)-5-((methylthio)methyl)tetrahydrofuran-3,4-diol, 5'-Methylthioadenosine
A191757-1MG
A191757-250MG
A191757-1G
A191757-50MG
A191757-100MG
103792-346EA
36
USD
103792-346
103799-458
103816-398
76825-294
76825-296
77084-866
77084-868
77683-501
5’-Deoxy-5’-methylthioadenosine 97%
5’-Deoxy-5’-methylthioadenosine
Precaución: For research use only.
Formula:
C₁₁H₁₅N₅O₃S MW: 297.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00010533 CAS Number: 2457-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...