5-Phenylpyridin-2-ol 98%
Supplier: Ambeed
|
Warning
|
A197737-100MG
A197737-250MG
A197737-5G
A197737-1G
103816-962EA
116
USD
103816-962
76814-148
76814-150
76814-152
77302-568
5-Phenylpyridin-2-ol 98%
5-Phenylpyridin-2-ol
Precaución: for Research Use Only
Formula:
C₁₁H₉NO MW: 171.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11044359 CAS Number: 76053-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...