5-Phenylnicotinamide 95%

Supplier: Ambeed
Warning

A146639-250MG A146639-1G
103803-690EA 87 USD
103803-690 103841-784 76984-482 77317-118
5-Phenylnicotinamide 95%
5-Phenylnicotinamide

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O
MW: 198.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19688859
CAS Number: 10177-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR