5-Phenyl-1,3-cyclohexanedione 98%

Supplier: Ambeed
Warning

Synonyms: 5-Phenylcyclohexane-1,3-dione

A180810-100G A180810-10G A180810-1G A180810-5G
103807-562EA 9 USD
103807-562 103845-924 76769-546 76769-548 77083-430 77083-432
5-Phenyl-1,3-cyclohexanedione 98%
5-Phenyl-1,3-cyclohexanedione

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
Boiling Pt: 356.5 °C (760 mm Hg)
Melting Pt: 188 °C
Flash Pt: 133.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00051846
CAS Number: 493-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR