5-Phenyl-1H-pyrrole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A169369-100MG A169369-250MG A169369-1G
103806-380EA 345 USD
103806-380 103844-634 76998-166 76998-168 77300-976
5-Phenyl-1H-pyrrole-2-carboxylic acid 97%
5-Phenyl-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MW: 187.2 g/mol
Boiling Pt: 452.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02575359
CAS Number: 6636-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR