3-Phenylpyrazole-5-carboxylic acid hydrate 97%
Supplier: Ambeed
|
Warning
|
A156933-250MG
A156933-1G
A156933-5G
103804-940EA
34
USD
103804-940
103843-142
77011-014
77318-948
77318-950
3-Phenylpyrazole-5-carboxylic acid hydrate 97%
3-Phenylpyrazole-5-carboxylic acid hydrate
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂O₂ Boiling Pt: 496.5 °C (760 mm Hg) Melting Pt: 250 °C Flash Pt: 254.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD01248821 CAS Number: 5071-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...