5-Phenyl-1H-indole-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A185448-100MG A185448-250MG A185448-1G A185448-5G
103815-854EA 215 USD
103815-854 103846-422 77248-400 77248-402 77301-976 77301-978
5-Phenyl-1H-indole-3-carbaldehyde 97%
5-Phenyl-1H-indole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₁NO
MW: 221.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06801952
CAS Number: 141835-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR