5-Phenyl-1H-imidazole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A128514-100MG A128514-250MG A128514-50MG
103801-148EA 760 USD
103801-148 76811-066 76811-068 76811-070
5-Phenyl-1H-imidazole-2-carboxylic acid 98%
5-Phenyl-1H-imidazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14583759
CAS Number: 41270-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR