5-Oxotetrahydrofuran-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A156433-100G
A156433-10G
A156433-250MG
A156433-1G
A156433-25G
A156433-5G
103804-880EA
7
USD
103804-880
103843-082
76783-606
76783-608
76783-610
77318-856
77318-858
77318-860
5-Oxotetrahydrofuran-2-carboxylic acid 95%
5-Oxotetrahydrofuran-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₆O₄ Boiling Pt: 384.2 °C (760 mm Hg) Melting Pt: 47…48 °C Flash Pt: 184.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066678 CAS Number: 4344-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...