4-Benzoylbutyric acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Oxo-5-phenylvaleric Acid, 5-Oxo-5-phenylpentanoic Acid, 5-Oxo-5-phenylpentanoic acid

A103242-5G A103242-10G
103822-214EA 17 USD
103822-214 77309-096 77570-190
4-Benzoylbutyric acid 98%
4-Benzoylbutyric acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 380.8 °C (760 mm Hg)
Melting Pt: 126…129 °C
Flash Pt: 198.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00004411
CAS Number: 1501-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR