5-Nitroquinoxaline 95%

Supplier: Ambeed
Warning

A599476-100MG A599476-250MG
103828-972EA 100 USD
103828-972 76775-178 76775-180
5-Nitroquinoxaline 95%
5-Nitroquinoxaline

Precaución: for Research Use Only

Formula: C₈H₅N₃O₂
Boiling Pt: 328.8 °C (760 mm Hg)
Flash Pt: 152.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01684951
CAS Number: 18514-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR