3-Nitro-6-pyridinecarboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-Nitropicolinaldehyde

A308170-250MG A308170-1G A308170-5G
103814-546EA 32 USD
103814-546 103835-722 77290-122 77305-978 77305-980
3-Nitro-6-pyridinecarboxaldehyde 98%
3-Nitro-6-pyridinecarboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₄N₂O₃
Boiling Pt: 298.7 °C (760 mm Hg)
Melting Pt: 54…57 °C
Flash Pt: 134.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368185
CAS Number: 35969-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR