5-Nitronicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Nitropyridin-3-ylcarboxylic acid, 5-Nitropyridine-3-carboxylic acid, 3-Carboxy-5-nitropyridine

A192645-1G A192645-5G A192645-10G A192645-250MG
103816-470EA 56 USD
103816-470 103847-080 77084-952 77084-954 77274-498 77274-500
5-Nitronicotinic acid 97%
5-Nitronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄N₂O₄
MW: 168.107 g/mol
Boiling Pt: 369.5 °C (760 mm Hg)
Melting Pt: 171…173 °C
Flash Pt: 177.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04038690
CAS Number: 2047-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR