5-Nitroindole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Nitro-1H-indole
A170191-25G
A170191-100G
A170191-10G
A170191-500G
A170191-5G
103844-724EA
13
USD
103844-724
77081-834
77273-676
77273-678
77273-680
77301-026
5-Nitroindole 97%
5-Nitroindole
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Boiling Pt: 362.6 °C (760 mm Hg) Melting Pt: 141…143 °C Flash Pt: 173.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005673 CAS Number: 6146-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...