5-Nitropicolinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Nitro-2-pyridinecarboxylic acid, 5-Nitropyridine-2-carboxylic acid, 5-Nitropicolinic acid

A137453-100G A137453-10G A137453-250MG A137453-1G A137453-25G A137453-5G
103802-458EA 6 USD
103802-458 76996-722 76996-724 76996-726 77315-416 77315-418 77315-420
5-Nitropicolinic acid 98%
5-Nitropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
Boiling Pt: 389.8 °C (760 mm Hg)
Melting Pt: 218…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114182
CAS Number: 30651-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR