5-Nitro-1H-pyrazolo[3,4-b]pyridine 95%

Supplier: Ambeed
Warning

A239289-100MG A239289-250MG A239289-1G
103811-276EA 44 USD
103811-276 103851-074 76976-400 76976-402 77304-152
5-Nitro-1H-pyrazolo[3,4-b]pyridine 95%
5-Nitro-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄N₄O₂
Boiling Pt: 385.6 °C (760 mm Hg)
Melting Pt: 212…213 °C
Storage Temperature: Ambient
MDL Number: MFCD11215386
CAS Number: 63572-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR