5-Nitro-1H-indole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A485898-100MG A485898-250MG A485898-1G A485898-5G
103828-174EA 33 USD
103828-174 103839-248 77265-552 77265-554 77307-562 77307-564
5-Nitro-1H-indole-3-carboxylic acid 97%
5-Nitro-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₄
MW: 206.16 g/mol
Boiling Pt: 520.8 °C (760 mm Hg)
Melting Pt: 254…258 °C
Flash Pt: 268.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06203492
CAS Number: 6958-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR