5-Methyluridine 97%

Supplier: Ambeed
Warning

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

A111045-100G A111045-10G A111045-25G A111045-500G
103808-060EA 257 USD
103808-060 103823-592 77270-892 77270-894 77270-896 77270-898
5-Methyluridine 97%
5-Methyluridine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₆
MW: 258.23 g/mol
Melting Pt: 183…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00006535
CAS Number: 1463-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR