5-Methyl-2-thiophenecarbonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 5-Methylthiophene-2-carbonitrile, 2-Cyano-5-methylthiophene

A191577-10G A191577-1G A191577-25G A191577-5G
103816-384EA 9 USD
103816-384 103846-986 76769-890 77084-826 77084-828 77084-830
5-Methyl-2-thiophenecarbonitrile 95%
5-Methyl-2-thiophenecarbonitrile

Precaución: for Research Use Only

Formula: C₆H₅NS
MW: 123.1756 g/mol
Boiling Pt: 219 °C (760 mm Hg)
Flash Pt: 86.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01434334
CAS Number: 72835-25-7
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR