5-Methylthiazole 98%

Supplier: Ambeed
Danger

A185257-10G A185257-5G
103815-836EA 9 USD
103815-836 103846-404 76769-710 77084-040
5-Methylthiazole 98%
5-Methylthiazole

Precaución: for Research Use Only

Formula: C₄H₅NS
MW: 99.16 g/mol
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00005335
CAS Number: 3581-89-3
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR