6-Hydroxy-m-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methylsalicylic acid, 2,5-Cresotic acid

A105323-10G A105323-5G A105323-100G
103822-596EA 7 USD
103822-596 77245-368 77309-466 77745-798
6-Hydroxy-m-toluic acid 98%
6-Hydroxy-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
Melting Pt: 150…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002461
CAS Number: 89-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR