5-Methylquinoxaline 98%

Supplier: Ambeed
Warning

A114281-100G A114281-10G A114281-25G A114281-5G A114281-500G
103824-224EA 9 USD
103824-224 77108-190 77108-192 77108-194 77311-034 77549-222
5-Methylquinoxaline 98%
5-Methylquinoxaline

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18
Boiling Pt: 244.8 °C (760 mm Hg)
Melting Pt: 20…21 °C
Storage Temperature: Ambient
MDL Number: MFCD00012335
CAS Number: 13708-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR