5-Methylpyrimidin-2-amine 98%

Supplier: Ambeed
Warning

A154541-10G A154541-1G
103804-652EA 23 USD
103804-652 103842-842 77273-094 77318-572
5-Methylpyrimidin-2-amine 98%
5-Methylpyrimidin-2-amine

Precaución: for Research Use Only

Formula: C5H7N3
MW: 109.1300 g/mol
Boiling Pt: 283.4 °C (760 mm Hg)
Melting Pt: 192…193 °C
Flash Pt: 149.873 °C
Storage Temperature: Ambient
MDL Number: MFCD00463477
CAS Number: 50840-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR