5-Methylpyridin-2-ol 97%

Supplier: Ambeed
Warning

A197592-10G A197592-25G A197592-5G A197592-100G
103847-568EA 7 USD
103847-568 76873-280 77085-584 77085-586 77883-433
5-Methylpyridin-2-ol 97%
5-Methylpyridin-2-ol

Precaución: for Research Use Only

Formula: C₆H₇NO
Boiling Pt: 304.2 °C (760 mm Hg)
Melting Pt: 183…187 °C
Flash Pt: 170.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03092888
CAS Number: 91914-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR