5-Methylpyrazin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-methylpyrazine

A165224-10G A165224-250MG A165224-1G A165224-25G A165224-5G A165224-100G
103805-948EA 14 USD
103805-948 103844-206 77163-328 77163-330 77320-400 77320-402 77320-404 77683-357
5-Methylpyrazin-2-amine 98%
5-Methylpyrazin-2-amine

Precaución: For research use only.

Formula: C5H7N3
MW: 109.13 g/mol
Boiling Pt: 244.3 °C (760 mm Hg)
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD08437664
CAS Number: 5521-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR