5-Methylpicolinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methylpyridine-2-carboxylic acid

A229562-10G A229562-250MG A229562-1G A229562-25G A229562-5G
103819-480EA 18 USD
103819-480 103850-380 76771-098 76771-100 77089-224 77089-226 77089-228
5-Methylpicolinic acid 98%
5-Methylpicolinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.1360 g/mol
Boiling Pt: 317 °C (760 mm Hg)
Flash Pt: 145.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01704358
CAS Number: 4434-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR