2-Formyl-5-picoline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Methylpyridine-2-carbaldehyde, 5-Methylpicolinaldehyde
A227613-1G
A227613-5G
A227613-100G
A227613-10G
103819-330EA
8
USD
103819-330
103850-238
77089-010
77089-012
77098-152
77098-154
2-Formyl-5-picoline 98%
2-Formyl-5-picoline
Precaución: for Research Use Only
Formula:
C₇H₇NO MW: 121.1400 g/mol Flash Pt: 84.245 °C Storage Temperature: Ambient |
MDL Number:
MFCD07367942 CAS Number: 4985-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...