5-Methyloxazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Oxazolecarboxylicacid, 5-methyl-, 5-Methyl-1,3-oxazole-4-carboxylic acid, 4-Carboxy-5-methyl-1,3-oxazole

A132719-10G A132719-250MG A132719-5G A132719-1G
103801-774EA 16 USD
103801-774 76767-304 76767-306 77298-576 77314-360
5-Methyloxazole-4-carboxylic acid 98%
5-Methyloxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₃
MW: 127.1 g/mol
Boiling Pt: 274.4 °C (760 mm Hg)
Flash Pt: 120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056309
CAS Number: 103879-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR