5-Methylisoxazole-4-boronic Acid Pinacol Ester 96%

Supplier: Ambeed
Warning

A101507-100MG A101507-250MG A101507-50MG A101507-5G
103796-006EA 305 USD
103796-006 76869-160 76869-162 76869-164 77882-874
5-Methylisoxazole-4-boronic Acid Pinacol Ester 96%
5-Methylisoxazole-4-boronic Acid Pinacol Ester

Precaución: for Research Use Only

Formula: C₁₀H₁₆BNO₃
MW: 209.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17677234
CAS Number: 1346808-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR