5-Methylisoxazole-3-carbonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 3-Cyano-5-methylisoxazole

A406931-10G A406931-5G
103827-146EA 27 USD
103827-146 103838-184 76773-762 76841-672
5-Methylisoxazole-3-carbonitrile 95%
5-Methylisoxazole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄N₂O
Boiling Pt: 249.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09064940
CAS Number: 57351-99-2
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR