3-Amino-5-methylisoxazole 97%

Supplier: Ambeed
Warning

A241010-100G A241010-10G A241010-25G A241010-5G A241010-500G
103851-194EA 10 USD
103851-194 77000-434 77000-436 77000-438 77090-350 77672-152
3-Amino-5-methylisoxazole 97%
3-Amino-5-methylisoxazole

Precaución: for Research Use Only

Formula: C₄H₆N₂O
Melting Pt: 59…61 °C
Flash Pt: 96.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00003155
CAS Number: 1072-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR