5-Methylisophthalic acid 97%
Supplier: Ambeed
|
Warning
|
A129962-100G
A129962-10G
A129962-500G
A129962-5G
A129962-25G
76972-014EA
52
USD
76972-014
76972-016
76972-018
77298-374
77313-774
5-Methylisophthalic acid 97%
5-Methylisophthalic acid
Precaución: for Research Use Only
Formula:
C₉H₈O₄ Boiling Pt: 408.7 °C (760 mm Hg) Melting Pt: 299…303 °C Flash Pt: 215.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013986 CAS Number: 499-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...