5-Methylisatin 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methylindoline-2,3-dione, 5-Methyl-1H-indole-2,3-dione

A227448-100G A227448-10G A227448-500G A227448-25G A227448-5G
103850-226EA 5 USD
103850-226 76692-000 76692-002 76692-004 77088-996 77088-998
5-Methylisatin 98%
5-Methylisatin

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00005721
CAS Number: 608-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR