5-Methylindole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-1H-indole

A210366-100G A210366-10G A210366-250MG A210366-1G A210366-25G A210366-5G
103817-998EA 14 USD
103817-998 103848-692 76691-434 76691-436 76691-438 77087-064 77087-066 77087-068
5-Methylindole 98%
5-Methylindole

Precaución: for Research Use Only

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 269 °C (760 mm Hg)
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00005680
CAS Number: 614-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR