5-Methylcytidine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one

A100251-250MG A100251-1G A100251-5G A100251-10G A100251-25G A100251-100G
103795-318EA 19 USD
103795-318 103821-028 76686-976 77296-610 77296-612 77599-466 77599-468 77623-802
5-Methylcytidine 97%
5-Methylcytidine

Precaución: for Research Use Only

Formula: C₁₀H₁₅N₃O₅
MW: 257.25 g/mol
Boiling Pt: 537.5 °C (760 mm Hg)
Melting Pt: 212…215 °C
Flash Pt: 278.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00006548
CAS Number: 2140-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR