3,5-Dihydroxytoluene monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methylresorcinol monohydrate, Orcinol monohydrate, 5-Methylbenzene-1,3-diol hydrate

A105327-100G A105327-10G A105327-25G A105327-5G
103822-598EA 13 USD
103822-598 77107-532 77107-534 77107-536 77309-468
3,5-Dihydroxytoluene monohydrate 98%
3,5-Dihydroxytoluene monohydrate

Precaución: for Research Use Only

Formula: C₇H₈O₂·1H₂O
Boiling Pt: 290 °C (760 mm Hg)
Melting Pt: 56…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00149092
CAS Number: 6153-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR